Assay Design
Gene Transcripts
- Gene Symbol
- Mcr
- Entrez Gene ID
-
44071
- Gene Name
- Macroglobulin complement-related
- Gene Aliases
-
CG7586, DmTEP6, Dmel\CG7586, Dmel_CG7586, E(br)155, TEP6, TEP6mel, Tep6, TepVI, alpha-2-Mrp, anon-28DE, dTEPVI, luf, mcr
- Chromosome Location
-
Chr.2L: 8074117 - 8082982 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0267488
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079949.3 |
NP_524688.1 |
4 - 5 |
4969 |
102 |
GenBank mRNA |
AY058526.1 |
- |
4 - 5 |
4935 |
102 |
View Full Product Details