Assay Design
Gene Transcripts
- Gene Symbol
- DCTN2-p50
- Entrez Gene ID
-
44086
- Gene Name
- Dynactin 2, p50 subunit
- Gene Aliases
-
CG8269, DCTN2, DMN, Dmel\CG8269, Dmel_CG8269, Dmn, Dynamitin, P50, dDyn, dmn, dyna, l(2)k16109, l(2)k16218, p50, p50/Dmn, p50/dynamitin, p50[Dmn]
- Chromosome Location
-
Chr.2R: 8891832 - 8893339 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0021825
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079951.3 |
NP_524690.1 |
2 - 3 |
1085 |
61 |
GenBank mRNA |
AY061092.1 |
- |
2 - 3 |
1062 |
61 |
View Full Product Details