Assay Design



Gene Transcripts

Gene Symbol
RabX1
Entrez Gene ID
44172
Gene Name
CG3870-PA
Gene Aliases
AAF47018, CG3870, Chrw, DmRabX1, Dmel\CG3870, Dmel_CG3870, RAB5c, chr, chrw, l(2)k06908, rabX1
Chromosome Location
Chr.2R: 23519839 - 23523613 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015372
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079974.3 NP_524713.1 2 - 3 518 59
GenBank mRNA AF209706.1 - 2 - 3 486 59
AY122200.1 - 2 - 3 478 59

View Full Product Details