Assay Design



Gene Transcripts

Gene Symbol
Sr-CII
Entrez Gene ID
44219
Gene Name
Scavenger receptor class C, type II
Gene Aliases
CG8856, CT25430, Dmel\CG8856, Dmel_CG8856, SR-CII, dSR-CII
Chromosome Location
Chr.2R: 12208893 - 12212155 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020377
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201979.2 NP_001188908.1 3 - 4 1186 63
NM_079981.3 NP_524720.2 2 - 3 860 63

View Full Product Details