Assay Design



Gene Transcripts

Gene Symbol
SoxN
Entrez Gene ID
44275
Gene Name
SoxNeuro
Gene Aliases
CG18024, DM64, Dmel\CG18024, Dmel_CG18024, SOX29F, SOXB1, Sox B1, Sry-rDM64, soxN
Chromosome Location
Chr.2L: 8825645 - 8829670 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029123
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273340.2 NP_001260269.1 1 - 1 3964 69
NM_079996.3 NP_524735.1 1 - 1 3964 69
GenBank mRNA BT001514.1 - 1 - 1 3127 69

View Full Product Details