Assay Design
Gene Transcripts
- Gene Symbol
- NiPp1
- Entrez Gene ID
-
44748
- Gene Name
- Nuclear inhibitor of Protein phosphatase 1
- Gene Aliases
-
CG8980, Dmel\CG8980, Dmel_CG8980, NIPP-1Dm, NIPP1, NIPP1Dm, NiPp-1, nipp1
- Chromosome Location
-
Chr.2R: 17003563 - 17005149 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026402
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137333.4 |
NP_611177.1 |
3 - 4 |
546 |
61 |
View Full Product Details