Assay Design



Gene Transcripts

Gene Symbol
NiPp1
Entrez Gene ID
44748
Gene Name
Nuclear inhibitor of Protein phosphatase 1
Gene Aliases
CG8980, Dmel\CG8980, Dmel_CG8980, NIPP-1Dm, NIPP1, NIPP1Dm, NiPp-1, nipp1
Chromosome Location
Chr.2R: 17003563 - 17005149 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026402
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137333.4 NP_611177.1 4 - 5 1126 63
GenBank mRNA AJ427611.1 - 4 - 5 63
AY069781.1 - 4 - 5 1066 63

View Full Product Details