Assay Design



Gene Transcripts

Gene Symbol
swi2
Entrez Gene ID
37010
Gene Name
CG14485-PA
Gene Aliases
CG14485, Dmel\CG14485, Dmel_CG14485
Chromosome Location
Chr.2R: 17690275 - 17695353 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034262
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137403.4 NP_611247.3 3 - 4 1042 72
GenBank mRNA AY084149.1 - 3 - 4 1040 72

View Full Product Details