Assay Design
Gene Transcripts
- Gene Symbol
- swi2
- Entrez Gene ID
-
37010
- Gene Name
- CG14485-PA
- Gene Aliases
-
CG14485, Dmel\CG14485, Dmel_CG14485
- Chromosome Location
-
Chr.2R: 17690275 - 17695353 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034262
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137403.4 |
NP_611247.3 |
4 - 5 |
1217 |
66 |
GenBank mRNA |
AY084149.1 |
- |
4 - 5 |
1215 |
66 |
View Full Product Details