Assay Design
Gene Transcripts
- Gene Symbol
- Oseg6
- Entrez Gene ID
-
37245
- Gene Name
- Outer segment 6
- Gene Aliases
-
CG11237, Dmel\CG11237, Dmel_CG11237, IFT144, OSEG, OSEG6, WDR19, Wdr19, oseg6
- Chromosome Location
-
Chr.2R: 19487046 - 19491846 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034452
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137582.3 |
NP_611426.1 |
6 - 7 |
2650 |
59 |
View Full Product Details