Assay Design
Gene Transcripts
- Gene Symbol
- Obp56b
- Entrez Gene ID
-
37265
- Gene Name
- Odorant-binding protein 56b
- Gene Aliases
-
56b, CG15129, CG30129, DmelObp56b, Dmel\CG30129, Dmel_CG30129, OBP56b
- Chromosome Location
-
Chr.2R: 19698714 - 19699346 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0046880
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137599.2 |
NP_611443.1 |
1 - 2 |
93 |
74 |
View Full Product Details