Assay Design
Gene Transcripts
- Gene Symbol
- ppk6
- Entrez Gene ID
-
37286
- Gene Name
- pickpocket 6
- Gene Aliases
-
CG11209, Dmel\CG11209, Dmel_CG11209, PPK6
- Chromosome Location
-
Chr.2R: 20245760 - 20247624 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034489
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137617.4 |
NP_611461.3 |
1 - 2 |
515 |
58 |
GenBank mRNA |
AY226539.1 |
- |
1 - 2 |
398 |
58 |
View Full Product Details