Assay Design
Gene Transcripts
- Gene Symbol
- maf-S
- Entrez Gene ID
-
37336
- Gene Name
- CG9954-PA
- Gene Aliases
-
CG9954, DM19, DM31, Dmel\CG9954, Dmel_CG9954, MAF-S, Maf, Maf-S, MafS, anon-EST:Posey173, maf2, mafS
- Chromosome Location
-
Chr.2R: 20640976 - 20641668 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034534
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137656.4 |
NP_611500.1 |
1 - 2 |
147 |
70 |
GenBank mRNA |
BT023717.1 |
- |
1 - 2 |
152 |
70 |
View Full Product Details