Assay Design



Gene Transcripts

Gene Symbol
Rgk3
Entrez Gene ID
5740446
Gene Name
Rad, Gem/Kir family member 3
Gene Aliases
CG15660, CG15663, CG15664, CG33949, CG34397, Dmel\CG34397, Dmel_CG34397, RGK3, mdcds_15673, mdcds_15683
Chromosome Location
Chr.2R: 21255165 - 21280995 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085426
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032275.2 NP_001027446.2 8 - 9 2597 58
NM_001103925.3 NP_001097395.1 8 - 9 1964 58
NM_001103926.4 NP_001097396.3 9 - 10 2717 58

View Full Product Details