Assay Design



Gene Transcripts

Gene Symbol
Paf-AHalpha
Entrez Gene ID
32529
Gene Name
Platelet-activating factor acetylhydrolase alpha
Gene Aliases
AAC83820, CG8962, Dmaalpha[[PAFAH]], Dmel\CG8962, Dmel_CG8962, PAFAHalpha
Chromosome Location
Chr.X: 15784009 - 15785583 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025809
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080350.4 NP_525089.2 2 - 3 850 76
NM_167461.2 NP_727895.1 1 - 2 922 76
GenBank mRNA AY069389.1 - 1 - 2 839 76

View Full Product Details