Assay Design



Gene Transcripts

Gene Symbol
par-6
Entrez Gene ID
32752
Gene Name
CG5884-PA
Gene Aliases
CG5884, DPar-6, DPar6, Dm-Par-6, Dm-Par6, DmPAR-6, DmPar-6, DmPar6, Dmel\CG5884, Dmel_CG5884, Dmpar6, PAR-6, PAR6, Par, Par-6, Par6, dPAR-6, dPar-6, dmPar-6, par, par6
Chromosome Location
Chr.X: 17703352 - 17707096 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026192
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133010.5 NP_573238.1 1 - 2 376 62
NM_167575.2 NP_728094.1 1 - 2 376 62
GenBank mRNA AF070969.1 - 1 - 2 351 62
AY061369.1 - 1 - 2 341 62

View Full Product Details