Assay Design
Gene Transcripts
- Gene Symbol
- Cda4
- Entrez Gene ID
-
33144
- Gene Name
- Chitin deacetylase-like 4
- Gene Aliases
-
BcDNA:RE51076, CG12447, CG13238, CG32499, CG9557, CG9558, DmCDA4, Dmel\CG32499, Dmel_CG32499
- Chromosome Location
-
Chr.X: 22583634 - 22632710 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052499
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_167788.2 |
NP_728468.1 |
3 - 4 |
402 |
89 |
GenBank mRNA |
AY113493.1 |
- |
3 - 4 |
401 |
89 |
View Full Product Details