Assay Design



Gene Transcripts

Gene Symbol
Pdk1
Entrez Gene ID
38017
Gene Name
Phosphoinositide-dependent kinase 1
Gene Aliases
1210, CG1201, CG1210, DSTPK61, Dmel\CG1210, Dmel_CG1210, Dstpk61, PDK, PDK-1, PDK1, PDK1/Pk61C, PK61C, Pdpk1, Pk61C, Pk61C/PDK1, Pk61C|PDK1, Pk61c/PDK1, dPDK, dPDK-1, dPDK1, dSTPK61, pdk1, pdpk1, pk61c
Chromosome Location
Chr.3L: 129450 - 145895 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020386
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274254.1 NP_001261183.1 2 - 3 828 53
NM_080382.4 NP_525121.2 2 - 3 1108 53
NM_167789.2 NP_728470.1 2 - 3 466 53
NM_167790.3 NP_728471.2 2 - 3 828 53
NM_167791.3 NP_728472.1 2 - 3 633 53
NM_167792.3 NP_728473.1 3 - 4 818 53
GenBank mRNA BT004489.1 - 2 - 3 1083 53
BT133220.1 - 2 - 3 711 53

View Full Product Details