Assay Design



Gene Transcripts

Gene Symbol
ttm50
Entrez Gene ID
31266
Gene Name
tiny tim 50
Gene Aliases
17944223, CG2713, Dmel\CG2713, Dmel_CG2713, EG:95B7.3, Gp99, Ttm50, i238, i90
Chromosome Location
Chr.X: 2730077 - 2732190 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0250874
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316628.1 NP_001303557.1 1 - 2 406 69
NM_130671.3 NP_570027.1 1 - 2 406 69
GenBank mRNA AY070535.1 - 1 - 2 382 69

View Full Product Details