Assay Design
Gene Transcripts
- Gene Symbol
- ppk12
- Entrez Gene ID
-
37566
- Gene Name
- pickpocket 12
- Gene Aliases
-
CG10972, Dmel\CG10972, Dmel_CG10972, PPK12
- Chromosome Location
-
Chr.2R: 22427479 - 22429584 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034730
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137828.1 |
NP_611672.1 |
4 - 5 |
753 |
84 |
GenBank mRNA |
AY226543.1 |
- |
4 - 5 |
753 |
84 |
View Full Product Details