Assay Design
Gene Transcripts
- Gene Symbol
- bonsai
- Entrez Gene ID
-
37587
- Gene Name
- CG4207-PA
- Gene Aliases
-
CG4207, DmMRPS15, Dmel\CG4207, Dmel_CG4207, MRP-S15, mRpS15
- Chromosome Location
-
Chr.2R: 22637770 - 22638868 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026261
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137847.3 |
NP_611691.1 |
1 - 2 |
291 |
75 |
GenBank mRNA |
AY060712.1 |
- |
1 - 2 |
260 |
75 |
View Full Product Details