Assay Design
Gene Transcripts
- Gene Symbol
- Obp58b
- Entrez Gene ID
-
37607
- Gene Name
- Odorant-binding protein 58b
- Gene Aliases
-
C13518, CG13518, DmelObp58b, Dmel\CG13518, Dmel_CG13518
- Chromosome Location
-
Chr.2R: 22704704 - 22705639 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034768
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137865.3 |
NP_611709.1 |
2 - 3 |
405 |
88 |
View Full Product Details