Assay Design



Gene Transcripts

Gene Symbol
asrij
Entrez Gene ID
37637
Gene Name
CG13533-PA
Gene Aliases
Asrij, CG13533, Dmasrij, Dmel\CG13533, Dmel_CG13533, FBgn 34793, arj
Chromosome Location
Chr.2R: 22886398 - 22887578 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034793
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274214.1 NP_001261143.1 1 - 2 146 79
NM_137889.3 NP_611733.1 1 - 2 309 79
GenBank mRNA AY089314.1 - 1 - 2 309 79

View Full Product Details