Assay Design
Gene Transcripts
- Gene Symbol
- pita
- Entrez Gene ID
-
37730
- Gene Name
- CG3941-PA
- Gene Aliases
-
CG3941, Dmel\CG3941, Dmel_CG3941, PITA, Pita, spdk
- Chromosome Location
-
Chr.2R: 23548967 - 23554712 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034878
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137962.3 |
NP_611806.3 |
1 - 2 |
458 |
62 |
GenBank mRNA |
AY061206.1 |
- |
1 - 2 |
458 |
62 |
View Full Product Details