Assay Design
Gene Transcripts
- Gene Symbol
- shu
- Entrez Gene ID
-
45360
- Gene Name
- shutdown
- Gene Aliases
-
CG4735, Dmel\CG4735, Dmel_CG4735, Shu
- Chromosome Location
-
Chr.2R: 23841461 - 23843286 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003401
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137993.4 |
NP_611837.1 |
2 - 3 |
900 |
73 |
GenBank mRNA |
AY058554.1 |
- |
2 - 3 |
870 |
73 |
View Full Product Details