Assay Design



Gene Transcripts

Gene Symbol
ocm
Entrez Gene ID
37874
Gene Name
over compensating males
Gene Aliases
CG3363, Dmel\CG3363, Dmel_CG3363, Mga, Mod(GMroX1)A
Chromosome Location
Chr.2R: 24186703 - 24195290 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266083
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202069.1 NP_001188998.1 3 - 4 2499 68
NM_001299902.1 NP_001286831.1 2 - 3 2405 68
NM_138050.4 NP_611894.1 3 - 4 2672 68

View Full Product Details