Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272447.2 | NP_001259376.1 | 3 - 4 | 3111 | 91 |
NM_001272449.1 | NP_001259378.1 | 4 - 5 | 3549 | 91 | |
NM_001369966.1 | NP_001356936.1 | 4 - 5 | 3413 | 91 | |
NM_143765.4 | NP_652022.1 | 4 - 5 | 3413 | 91 | |
GenBank mRNA | BT025212.1 | - | 4 - 5 | 3416 | 91 |