Assay Design
Gene Transcripts
- Gene Symbol
- gry
- Entrez Gene ID
-
38391
- Gene Name
- gryzun
- Gene Aliases
-
CG17569, Dmel\CG17569, Dmel_CG17569, TRAPPC11, gryz, l(3)63Bd
- Chromosome Location
-
Chr.3L: 3199090 - 3211348 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0286567
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139528.3 |
NP_647785.3 |
6 - 7 |
2517 |
59 |
View Full Product Details