Assay Design



Gene Transcripts

Gene Symbol
Ets65A
Entrez Gene ID
38700
Gene Name
Ets at 65A
Gene Aliases
CG13294, CG6882, CG7018, D-Ets-3, D-ets-3, Dmel\CG7018, Dmel_CG7018, ETS3, Ets, Ets3, ets65A
Chromosome Location
Chr.3L: 6097617 - 6123913 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005658
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144422.2 NP_001137894.2 3 - 4 1527 79
NM_001316444.1 NP_001303373.1 3 - 4 1527 79
NM_001316445.1 NP_001303374.1 3 - 4 1527 79
NM_168156.2 NP_729142.1 3 - 4 1527 79

View Full Product Details