Assay Design



Gene Transcripts

Gene Symbol
shf
Entrez Gene ID
31617
Gene Name
shifted
Gene Aliases
CG3134, CG3135, CT10514, D-Wif, DmWIF, DmWif, Dmel\CG3135, Dmel_CG3135, Shf, shf-ov
Chromosome Location
Chr.X: 6771463 - 6779659 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003390
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298034.1 NP_001284963.1 2 - 3 656 84
NM_001298035.1 NP_001284964.1 2 - 3 656 84
NM_001298036.1 NP_001284965.1 2 - 3 656 84
NM_132121.3 NP_572349.1 2 - 3 656 84
GenBank mRNA AY051622.1 - 2 - 3 656 84

View Full Product Details