Assay Design



Gene Transcripts

Gene Symbol
snz
Entrez Gene ID
31704
Gene Name
snazarus
Gene Aliases
CG1514, DmSNX25, Dmel\CG1514, Dmel_CG1514, SNZ, Snz
Chromosome Location
Chr.X: 7912089 - 7918444 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029976
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298064.1 NP_001284993.1 6 - 7 2020 69
NM_132193.2 NP_572421.1 6 - 7 2020 69
GenBank mRNA BT150246.1 - 6 - 7 1787 69

View Full Product Details