Assay Design
Gene Transcripts
- Gene Symbol
- Eps-15
- Entrez Gene ID
-
37961
- Gene Name
- Epidermal growth factor receptor pathway substrate clone 15
- Gene Aliases
-
Aa2/2, CG16932, Dmel\CG16932, Dmel_CG16932, EH1, EPS-15, EPS15, Eps15, anon-WO0118547.85, eps-15, eps15, i31
- Chromosome Location
-
Chr.2R: 24768314 - 24775265 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035060
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138121.2 |
NP_611965.2 |
6 - 7 |
1834 |
64 |
View Full Product Details