Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038885.2 | NP_001033974.1 | 1 - 2 | 1003 | 67 |
NM_138139.4 | NP_611983.2 | 2 - 3 | 488 | 67 | |
GenBank mRNA | AY060353.1 | - | 1 - 2 | 992 | 67 |
BT011448.1 | - | 2 - 3 | 484 | 67 | |
KX531560.1 | - | 1 - 2 | 90 | 67 |