Assay Design



Gene Transcripts

Gene Symbol
mthl9
Entrez Gene ID
38056
Gene Name
methuselah-like 9
Gene Aliases
CG17084, Dmel\CG17084, Dmel_CG17084, Mth-9, Mthl9, anon-WO0170980.163, anon-WO0170980.164
Chromosome Location
Chr.3L: 329822 - 332309 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035131
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138185.4 NP_612029.1 3 - 4 1438 77
GenBank mRNA AY070598.1 - 3 - 4 1435 77
AY113401.1 - 3 - 4 1435 77

View Full Product Details