Assay Design



Gene Transcripts

Gene Symbol
RabX6
Entrez Gene ID
38084
Gene Name
CG12015-PA
Gene Aliases
CG12015, DmRabX6, Dmel\CG12015, Dmel_CG12015, rabX6, rabx6
Chromosome Location
Chr.3L: 690517 - 691951 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035155
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274290.1 NP_001261219.1 1 - 2 288 85
NM_138208.4 NP_612052.1 1 - 2 288 85
GenBank mRNA AY071647.1 - 1 - 2 286 85

View Full Product Details