Assay Design
Gene Transcripts
- Gene Symbol
- SA-2
- Entrez Gene ID
-
38167
- Gene Name
- Stromalin-2
- Gene Aliases
-
CG1, CG13916, Dmel\CG13916, Dmel_CG13916, SNM, anon-WO0109301.35, pf-1, snm
- Chromosome Location
-
Chr.3L: 1434336 - 1437872 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0043865
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138265.3 |
NP_612109.3 |
5 - 6 |
2637 |
95 |
View Full Product Details