Assay Design
Gene Transcripts
- Gene Symbol
- cue
- Entrez Gene ID
-
38174
- Gene Name
- cueball
- Gene Aliases
-
CG12086, CT5570, Dmel\CG12086, Dmel_CG12086, ms(3)02509
- Chromosome Location
-
Chr.3L: 1503226 - 1517271 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011204
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138269.3 |
NP_612113.2 |
1 - 2 |
355 |
85 |
GenBank mRNA |
AY061139.1 |
- |
1 - 2 |
322 |
85 |
View Full Product Details