Assay Design



Gene Transcripts

Gene Symbol
Rdl
Entrez Gene ID
39054
Gene Name
Resistant to dieldrin
Gene Aliases
CG10537, CT29555, DmRDL, DmRdl, Dmel\CG10537, Dmel_CG10537, GABA, GABA-R, GABA[[A]], GABA[[A]] receptor, GABA[[A]]-R, GABA[[A]]R, GABAr, LCCH1, RDL, Rd1, gaba, rdl
Chromosome Location
Chr.3L: 9145838 - 9182149 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004244
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274688.1 NP_001261617.1 7 - 8 2403 63
NM_168321.3 NP_729461.1 7 - 8 2403 63
NM_168322.3 NP_729462.2 7 - 8 2499 63

View Full Product Details