Assay Design
Gene Transcripts
- Gene Symbol
- Rbfox1
- Entrez Gene ID
-
39198
- Gene Name
- RNA-binding Fox protein 1
- Gene Aliases
-
A2BP1, A2bp1, BcDNA:GM06073, CG14155, CG18441, CG32062, Dmel\CG32062, Dmel_CG32062, cg18441, dA2BP1, rbfox1
- Chromosome Location
-
Chr.3L: 10481412 - 10594012 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052062
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168412.4 |
NP_729614.2 |
11 - 12 |
2900 |
76 |
View Full Product Details