Assay Design



Gene Transcripts

Gene Symbol
Rbfox1
Entrez Gene ID
39198
Gene Name
RNA-binding Fox protein 1
Gene Aliases
A2BP1, A2bp1, BcDNA:GM06073, CG14155, CG18441, CG32062, Dmel\CG32062, Dmel_CG32062, cg18441, dA2BP1, rbfox1
Chromosome Location
Chr.3L: 10481412 - 10594012 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052062
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104102.4 NP_001097572.1 14 - 15 3062 68
NM_001104103.3 NP_001097573.1 11 - 12 2967 68
NM_001104104.3 NP_001097574.2 16 - 17 2840 68
NM_001259778.1 NP_001246707.1 16 - 17 3810 68
NM_001259779.1 NP_001246708.1 13 - 14 3498 68
NM_001274751.1 NP_001261680.1 13 - 14 2093 68
NM_001370003.1 NP_001356980.1 11 - 12 2520 68
NM_168412.4 NP_729614.2 13 - 14 3119 68
NM_168413.3 NP_729615.2 15 - 16 3215 68
GenBank mRNA BT010058.1 - 14 - 15 3034 68

View Full Product Details