Assay Design



Gene Transcripts

Gene Symbol
Rbfox1
Entrez Gene ID
39198
Gene Name
RNA-binding Fox protein 1
Gene Aliases
A2BP1, A2bp1, BcDNA:GM06073, CG14155, CG18441, CG32062, Dmel\CG32062, Dmel_CG32062, cg18441, dA2BP1, rbfox1
Chromosome Location
Chr.3L: 10481412 - 10594012 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052062
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104102.4 NP_001097572.1 8 - 9 1927 59
NM_001104103.3 NP_001097573.1 8 - 9 1912 59
NM_001259779.1 NP_001246708.1 8 - 9 2435 59
NM_168412.4 NP_729614.2 9 - 10 2110 59
GenBank mRNA BT010058.1 - 8 - 9 1899 59

View Full Product Details