Assay Design



Gene Transcripts

Gene Symbol
Mob2
Entrez Gene ID
39293
Gene Name
CG11711-PA
Gene Aliases
BEST:GH06341, BcDNA:GH13236, CG11711, CG11712, DMob2, Dmel\CG11711, Dmel_CG11711, Dmob1, Dmob2, Mob1, MobI, dmob2, gh06341, trezor
Chromosome Location
Chr.3L: 11522757 - 11562233 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259481
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274784.1 NP_001261713.1 2 - 3 499 75
NM_168462.2 NP_729716.1 2 - 3 450 75
GenBank mRNA BT124999.1 - 2 - 3 430 75

View Full Product Details