Assay Design



Gene Transcripts

Gene Symbol
rols
Entrez Gene ID
39368
Gene Name
rolling pebbles
Gene Aliases
ANTS, Ants, CG12277, CG17155, CG32096, CG5679, Dmel\CG32096, Dmel_CG32096, Rol/Ants, Rols, Rols6, Rols7, anon-WO0257455.19, ants, ants/rols, l(3)08232, l(3)68Fd, l(3)j2A6, rols\ants
Chromosome Location
Chr.3L: 12008225 - 12064859 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041096
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274807.1 NP_001261736.1 6 - 7 1901 56
NM_168487.2 NP_729778.1 6 - 7 2330 56
NM_168489.2 NP_729780.1 6 - 7 1887 56
NM_168490.2 NP_729781.1 6 - 7 1904 56
GenBank mRNA AF440568.1 - 5 - 6 1460 56

View Full Product Details