Assay Design



Gene Transcripts

Gene Symbol
Nc73EF
Entrez Gene ID
39899
Gene Name
Neural conserved at 73EF
Gene Aliases
141810_at, CG11661, DOGDH, Dmel\CG11661, Dmel_CG11661, E1, Nc73eF, anon-WO0118547.329, dOgdh
Chromosome Location
Chr.3L: 16954822 - 16968724 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010352
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104159.3 NP_001097629.1 2 - 3 208 88
NM_168701.5 NP_730223.1 2 - 3 208 88
NM_168702.2 NP_730224.1 2 - 3 169 88
NM_168703.5 NP_730225.1 2 - 3 211 88
NM_168704.4 NP_730226.3 3 - 4 556 88
NM_176340.3 NP_788518.1 2 - 3 162 88
NM_176341.3 NP_788519.1 2 - 3 162 88
NM_176342.5 NP_788520.2 2 - 3 186 88
GenBank mRNA BT003743.1 - 2 - 3 189 88
BT012466.1 - 2 - 3 149 88

View Full Product Details