Assay Design



Gene Transcripts

Gene Symbol
Nc73EF
Entrez Gene ID
39899
Gene Name
Neural conserved at 73EF
Gene Aliases
141810_at, CG11661, DOGDH, Dmel\CG11661, Dmel_CG11661, E1, Nc73eF, anon-WO0118547.329, dOgdh
Chromosome Location
Chr.3L: 16954822 - 16968724 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010352
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104159.3 NP_001097629.1 9 - 10 2885 74
NM_168701.5 NP_730223.1 9 - 10 2885 74
NM_168702.2 NP_730224.1 9 - 10 2846 74
NM_168703.5 NP_730225.1 9 - 10 2888 74
NM_168704.4 NP_730226.3 10 - 11 3233 74
NM_176340.3 NP_788518.1 9 - 10 2866 74
NM_176341.3 NP_788519.1 9 - 10 2839 74
NM_176342.5 NP_788520.2 9 - 10 2890 74
GenBank mRNA BT003743.1 - 9 - 10 2866 74
BT012466.1 - 9 - 10 2853 74

View Full Product Details