Assay Design



Gene Transcripts

Gene Symbol
ome
Entrez Gene ID
44297
Gene Name
omega
Gene Aliases
BEST:LD21715, CG17705, CG32143, CG32145, CG42280, CG9370, Dmel\CG42280, Dmel_CG42280, LD21715, Ome
Chromosome Location
Chr.3L: 14672839 - 14747868 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259175
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104136.2 NP_001097606.1 9 - 10 2970 64
NM_001144483.2 NP_001137955.1 8 - 9 3009 64
NM_001274915.1 NP_001261844.1 7 - 8 2163 64
NM_080000.4 NP_524739.1 8 - 9 2256 64
GenBank mRNA AY058514.1 - 8 - 9 2229 64
BT024224.1 - 8 - 9 2980 64
BT029957.1 - 9 - 10 2954 64

View Full Product Details