Assay Design
Gene Transcripts
- Gene Symbol
- msk
- Entrez Gene ID
-
44747
- Gene Name
- moleskin
- Gene Aliases
-
61, CG7935, CIP-61, DIM-7, Dim-7, Dim7, DmRanBP7, Dmel\CG7935, Dmel_CG7935, Imp7, Imp7/8, Msk, S(ls)2, dim-7, dim7
- Chromosome Location
-
Chr.3L: 8116179 - 8121579 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026252
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080041.3 |
NP_524780.1 |
3 - 4 |
1082 |
106 |
GenBank mRNA |
BT053682.1 |
- |
3 - 4 |
1083 |
106 |
View Full Product Details