Assay Design



Gene Transcripts

Gene Symbol
msk
Entrez Gene ID
44747
Gene Name
moleskin
Gene Aliases
61, CG7935, CIP-61, DIM-7, Dim-7, Dim7, DmRanBP7, Dmel\CG7935, Dmel_CG7935, Imp7, Imp7/8, Msk, S(ls)2, dim-7, dim7
Chromosome Location
Chr.3L: 8116179 - 8121579 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026252
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080041.3 NP_524780.1 5 - 6 1406 67
GenBank mRNA AF132299.1 - 5 - 6 1315 67
BT053682.1 - 5 - 6 1407 67

View Full Product Details