Assay Design
Gene Transcripts
- Gene Symbol
- Tango4
- Entrez Gene ID
-
32168
- Gene Name
- Transport and Golgi organization 4
- Gene Aliases
-
CG1796, Dmel\CG1796, Dmel_CG1796, TANGO4, tango4
- Chromosome Location
-
Chr.X: 12021068 - 12022746 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030365
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132550.3 |
NP_572778.1 |
1 - 2 |
216 |
71 |
GenBank mRNA |
AY051707.1 |
- |
1 - 2 |
176 |
71 |
View Full Product Details