Assay Design



Gene Transcripts

Gene Symbol
rad
Entrez Gene ID
32253
Gene Name
radish
Gene Aliases
CG15720, CG18646, CG42628, CG42629, CG4346, CG4368, CG44424, Dmel\CG44424, Dmel_CG44424, FBpp0073550, R5_X:12971309..12973132, Rad, Rsh, rsh
Chromosome Location
Chr.X: 12988615 - 13079070 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0265597
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298247.1 NP_001285176.1 4 - 5 1954 74
NM_001298248.1 NP_001285177.1 4 - 5 2178 74
NM_001298249.1 NP_001285178.1 4 - 5 1954 74
NM_001298250.1 NP_001285179.1 4 - 5 2178 74
NM_001298251.1 NP_001285180.1 4 - 5 1954 74

View Full Product Details